Order Entry
Italy
ContactUsLinkComponent
 
 
1-Adamantanecarbonyl chloride 97%
 
undefined
 
1-Adamantanecarbonyl chloride 97%

 

  • Appearance:
    White to off-white Crystalline solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    48 to 52 °C
  • Titration RCOCl:
    ≥96.0 %

 

 

Frequently Bought Together