5-Methylthiophene-2-carboxaldehyde 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13679-70-4
:
Formula:C₆H₆OS
MDL Number:MFCD00005434
:126.18 g/mol
Density:1.18 g/cm³
Flash Pt:87 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear yellow to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5800 to 1.5840 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together