Order Entry
Italy
ContactUsLinkComponent
1,2-Di(4-pyridyl)ethane 97%
Irritant
1,2-Di(4-pyridyl)ethane 97%
Certificates
undefined
Irritant
1,2-Di(4-pyridyl)ethane 97%

Specification Test Results

  • Appearance:
    White to light yellow to pink Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    107 to 113 °C
  • Titration with HClO4:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together