Order Entry
Italy
Orders LinkContactUsLinkComponent
Methyl 4-(1-hydroxyethyl)benzoate 90% tech.

Methyl 4-(1-hydroxyethyl)benzoate 90% tech.

Irritant
Methyl 4-(1-hydroxyethyl)benzoate 90% tech.
Certificates
Methyl 4-(1-hydroxyethyl)benzoate 90% tech.

2 Options Available Below

Product Details & Documents

CAS No.: 79322-76-2

Synonyms:  1-[4-(Methoxycarbonyl)phenyl]ethan-1-ol, 4-(Methoxycarbonyl)-α-methylbenzyl alcohol
  • CAS Number:79322-76-2
  • Formula:C₁₀H₁₂O₃
  • MDL Number:MFCD00216476
  • Molecular Weight:180.203 g/mol
  • Boiling Pt:148…152 °C (1 mmHg)
  • Flash Pt:>93 °C
  • Storage Temperature:Ambient

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to yellow Viscous liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5300 to 1.5360 (20°C, 589 nm)
  • GC:
    ≥88 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 67,40 €
    67,40 €
    1 * 1 g
       67,40 €

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 278,00 €
    278,00 €
    1 * 5 g
       278,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.