
Guanylurea sulphate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:591-01-5
:
Formula:C₂H₈N₄O₅S
MDL Number:MFCD00035565
Melting Pt:190 °C (dec.)
:200.17 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Water:≤8 % (K.F.)
- Titration with NaOH:≥97.5 % (On dry substance)
Specification Test Results
Frequently Bought Together