
Dodecylamine hydrochloride 99%
Catalog # ACRO409250100
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥98.5 %
Specification Test Results
About this item
:
Melting Pt:186…188 °C
:221.81 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:929-73-7
Formula:C₁₂H₂₇N
MDL Number:MFCD00042017