

Dimethylthiocarbamoyl chloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16420-13-6
:
Formula:C₃H₆ClNS
MDL Number:MFCD00004919
:123.60 g/mol
Boiling Pt:63…65 °C (.2mbar)
Melting Pt:42…43 °C
Flash Pt:98 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS08
:GHS05
Specification Test Results
- Appearance:Yellow Crystals or low melting crystalline solid
- Infrared spectrum:Conforms
- GC:≥98.5 %
- Appearance of solution:(10% in CHCl3) clear faint yellow to yellow
Specification Test Results
Frequently Bought Together