Order Entry
Italy
ContactUsLinkComponent
 
Flammable
Irritant
Health
3,6,9-Triethyl-3,6,9-trimethyl-1,4,7-triperoxonane 41% in aromatic free mineral spirit
Catalog # ACRO349941000
CAS Number:  
undefined
Flammable
Irritant
Health
3,6,9-Triethyl-3,6,9-trimethyl-1,4,7-triperoxonane 41% in aromatic free mineral spirit
Catalog # ACRO349941000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Appearance:
    Clear solution
  • Titration Iodometric:
    7.3 to 7.6 % (total active oxygen)
  • Color scale:
    ≤50 APHA (at 40°C)

Specification Test Results

About this item

CAS Number:24748-23-0
 :
Formula:C₁₂H₂₄O₆
MDL Number:MFCD02685709
 :264.318 g/mol
Melting Pt:< 10 °C
Density:0.875 g/cm³
Flash Pt:74 °C
UN:3105
ADR:5.2,
 :GHS02
 :GHS07
 :GHS08
Storage Temperature:Ambient