


2-(4-Chlorosulphonylphenyl)ethyltrimethoxysilane 50% in dichloromethane
:126519-89-9
:
:C₁₁H₁₇ClO₅SSi
:MFCD00054774
:324.847 g/mol
:>40 °C (760 mmHg)
:3265
:8,II
:GHS05
:GHS07
:GHS08
:Ambient
- Appearance:Clear yellow to light brown Solution
- Infrared spectrum:Conforms
- Assay:45 to 55 % solids
Frequently Bought Together