Order Entry
Italy
ContactUsLinkComponent
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide 98%
Corrosive
Irritant
Health
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide 98%
Certificates
undefined
Corrosive
Irritant
Health
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide 98%

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    92 to 97 °C
  • Titration Argentometric:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together