Order Entry
Italy
ContactUsLinkComponent
3-Amino-N,N-dimethyl-4-nitroaniline 97%
Irritant
3-Amino-N,N-dimethyl-4-nitroaniline 97%
Catalog # ACRO326220050
CAS Number:  
undefined
Irritant
3-Amino-N,N-dimethyl-4-nitroaniline 97%
Catalog # ACRO326220050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Yellow-brown Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    134 to 139 °C
  • GC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :181.195 g/mol
Melting Pt:134…139 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:2069-71-8
Formula:C₈H₁₁N₃O₂
MDL Number:MFCD01320677