

3-Bromopyruvic acid hydrate 98%
ACRO325690250
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:Beige to light brown Powder
- Infrared spectrum:Conforms
- Titration with NaOH:≥97.5 %
:
:79…82 °C
:Refrigerator
:3261
:8,II
:166.96 g/mol (anhy)
:GHS07
:GHS05
:206860-50-6
:1113-59-3
:C₃H₃BrO₃.xH₂O
:MFCD00149107