
Ethyl-3-aminopropanoate hydrochloride 98%
:4244-84-2
:
:C₅H₁₁NO₂·HCl
:MFCD00012909
:153.6 g/mol
:55…57 °C
:GHS07
:Ambient
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Titration Argentometric:97.5 to 102.5 %
- TLC:≥98.5 %
Frequently Bought Together