Tetrakis(1-pyrazolyl)borate potassium salt 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14782-58-2
:
Formula:C₁₂H₁₂BKN₈O₄
MDL Number:MFCD00800557
:382.188 g/mol
Melting Pt:253…254 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Titration with HClO4:≥94 %
Specification Test Results
Frequently Bought Together