Amberlite® XAD-1180 polymeric adsorbent
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9003-69-4
:97396-56-0
MDL Number:MFCD00145830
Flash Pt:298 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Beads
- Particle size:≤5.0 % (< 0.250 mm)
- Harmonic mean size:0.350 to 0.600 mm
- Uniformity coefficient:≤1.80
- Moisture holding capacity:61 to 67 %
Specification Test Results
Frequently Bought Together