Order Entry
Italy
ContactUsLinkComponent
trans-2-(4-Bromobenzoyl)-1-cyclohexanecarboxylic acid 98%
trans-2-(4-Bromobenzoyl)-1-cyclohexanecarboxylic acid 98%
Catalog # ACRO298690010
CAS Number:  
undefined
trans-2-(4-Bromobenzoyl)-1-cyclohexanecarboxylic acid 98%
Catalog # ACRO298690010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    163 to 169 °C
  • Titration with NaOH:
    ≥97.5 %

Specification Test Results

About this item

 :
 :311.175 g/mol
Melting Pt:163…169 °C
Storage Temperature:Ambient
CAS Number:212757-09-0
Formula:C₁₄H₁₅BrO₃
MDL Number:MFCD00800528