Order Entry
Italy
ContactUsLinkComponent
Ethyl-cis-2-amino-1-cyclopentane carboxylate hydrochloride 99%
Ethyl-cis-2-amino-1-cyclopentane carboxylate hydrochloride 99%
Catalog # ACRO298392500
CAS Number:  
undefined
Ethyl-cis-2-amino-1-cyclopentane carboxylate hydrochloride 99%
Catalog # ACRO298392500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    97 to 100 °C
  • Titration with HClO4:
    ≥98.5 %

Specification Test Results

About this item

 :
 :193.671 g/mol
Melting Pt:97…100 °C
Storage Temperature:Ambient
CAS Number:142547-15-7
Formula:C₈H₁₆ClNO₂
MDL Number:MFCD00800503