
1-Methyl-2-benzimidazolinone 98%
Catalog # ACRO276500050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to light beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:192 to 196 °C
- HPLC:≥97.5 %
Specification Test Results
About this item
:
:148.165 g/mol
Melting Pt:192…196 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1849-01-0
Formula:C₈H₈N₂O
MDL Number:MFCD00142654