Order Entry
Italy
ContactUsLinkComponent
1,2:5,6-Di-O-isopropylidene-α-D-allofuranose 98%
1,2:5,6-Di-O-isopropylidene-α-D-allofuranose 98%
  ACRO270960100
 :  
undefined
1,2:5,6-Di-O-isopropylidene-α-D-allofuranose 98%
  ACRO270960100
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

 

 

  • Appearance:
    White to off-white Crystalline powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    72 to 77 °C
  • GC:
    ≥97.5 %
  • Specific optical rotation:
    +33.0° to +39.0° (20 °C, 589 nm) (c=1, CHCl3)

 

 

 :
 :260.28 g/mol
 :73…77 °C
 :Refrigerator
 :2595-05-3
 :C₁₂H₂₀O₆
 :MFCD00135634