1,6-Anhydro-β-D-glucose 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:498-07-7
:
Formula:C₆H₁₀O₅
MDL Number:MFCD00063248
:162.14 g/mol
Melting Pt:170…180 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder or crystals
- Infrared spectrum:Conforms
- Water:≤1.0 % (K.F.)
- Solubility:(5 % in water) Clear colorless
- HPLC:≥99.0 % (ELSD)
- Specific optical rotation:-64° to -68° (20°C, 589 nm) (c=2, H2O)
Specification Test Results
Frequently Bought Together