3-Aminobicyclo[2.2.1]hept-5-ene-2-carboxylic acid 98%
Catalog # ACRO265690010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Beige to light grey Powder
- Infrared spectrum:Conforms
- Water:≤10.5 % (K.F.)
- Titration with HClO4:≥97.5 % (On dry substance)
Specification Test Results
About this item
:
:153.181 g/mol
Melting Pt:261…263 °C
Storage Temperature:Ambient
CAS Number:92511-32-5
Formula:C₈H₁₁NO₂