
trans-(+)-Chrysanthemic acid 99+%
Catalog # ACRO263635000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mg
- Seq:2725
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4760 to 1.4790 (20°C, 589 nm)
- GC:≥99.0 %
- Specific optical rotation:+13° to +15° (20°C, 589 nm) (c=2, ethanol)
Specification Test Results
About this item
:
:168.236 g/mol
Melting Pt:17…21 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:4638-92-0
Formula:C₁₀H₁₆O₂
MDL Number:MFCD01941572