
Isoguanine 98%
Catalog # ACRO241752500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mg
- Seq:2590
Specifications
Specification Test Results
- Appearance:Off-white to light beige Powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
Specification Test Results
About this item
:
:151.13 g/mol
Melting Pt:300 °C
:GHS07
Storage Temperature:Ambient
CAS Number:3373-53-3
Formula:C₅H₅N₅O
MDL Number:MFCD00142931