Sodium-5-oxopyrrolidine-2-carboxylate 50% (w/w) in water
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:54571-67-4
:
Formula:C₅H₆NNaO₃
MDL Number:MFCD00066984
Boiling Pt:109 °C
Density:1.27 g/cm³
:151.097 g/mol
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to light yellow Solution
- Water:47.5 to 52.5 % (K.F.)
- Heavy metals:≤20 ppm
- pH:6.8 to 7.4
- Titration with HClO4:47.5 to 52.5 %
- Arsenic (As):≤2 ppm
Specification Test Results
Frequently Bought Together