Order Entry
Italy
ContactUsLinkComponent
2'-Deoxyguanosine-5'-monophosphate, sodium salt hydrate 99%
2'-Deoxyguanosine-5'-monophosphate, sodium salt hydrate 99%
Certificates
undefined
2'-Deoxyguanosine-5'-monophosphate, sodium salt hydrate 99%

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤20 % (K.F.)
  • Solubility:
    (50 mg/ml in water) Clear colorless
  • HPLC:
    ≥98.5 %
  • Ratio of Absorbances:
    0.65 to 0.68 (A280/A260) c=1, water

Specification Test Results

Related Information

Frequently Bought Together