Order Entry
Italy
ContactUsLinkComponent
Anti-Myc Tag Rabbit Polyclonal Antibody (CF640R)
Anti-Myc Tag Rabbit Polyclonal Antibody (CF640R)
Catalog # 20439.
Supplier:  Biotium
Anti-Myc Tag Rabbit Polyclonal Antibody (CF640R)
Catalog # 20439.
Supplier:  Biotium

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    Myc Tag (EQKLISEEDL)
  • Antigen symbol:
    Myc Tag
  • Clonality:
    Polyclonal
  • Conjugation:
    CF® 640R
  • Host:
    Rabbit
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
  • Storage temperature:
    Store at –20 °C.
  • Concentration:
    1 mg/mL
  • Purification:
    Affinity purified
  • Size:
    100 μL
  • Pk:
    100 µl

Specifications

About this item

Polyclonal anti-MYC (EQKLISEEDL) reacts with human c- MYC tagged fusion proteins. The antibody recognizes the epitope tag fused to either the amino- or carboxy- termini of targeted proteins. Polyclonal anti-MYC (EQKLISEEDL) reacts with human c- MYC tagged fusion proteins. The antibody recognizes the epitope tag fused to either the amino- or carboxy- termini of targeted proteins. This conjugate is prepared by labeling polyclonal rabbit anti-MYC IgG (H+L) with CF™640R dye. CF™640R (Ex/Em 642/662 nm) is a novel far-red rhodamine-based dye spectrally similar to Cy®5 and Alexa Fluor® 647. CF™640R is much brighter than Cy®5 and at least as bright as Alexa Fluor® 647 when excited at 633 or 635 nm. However, the most important advantage of CF™640R is its exceptional photostability.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.

Type: Tag
Antigen: Myc Tag
Clonality: Polyclonal
Clone:
Conjugation: CF640R
Epitope:
Host: Rabbit
Isotype:
Reactivity: MYC tag