Order Entry
Italy
ContactUsLinkComponent
Anti-Myc Tag Rabbit Polyclonal Antibody (CF543)
Anti-Myc Tag Rabbit Polyclonal Antibody (CF543)
Catalog # 20437.
Supplier:  Biotium
Anti-Myc Tag Rabbit Polyclonal Antibody (CF543)
Catalog # 20437.
Supplier:  Biotium

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    Myc Tag (EQKLISEEDL)
  • Antigen symbol:
    Myc Tag
  • Clonality:
    Polyclonal
  • Conjugation:
    CF® 543
  • Host:
    Rabbit
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
  • Storage temperature:
    Store at –20 °C.
  • Concentration:
    1 mg/mL
  • Purification:
    Affinity purified
  • Size:
    100 μL
  • Pk:
    100 µl

Specifications

About this item

Polyclonal anti-MYC (EQKLISEEDL) reacts with human c- MYC tagged fusion proteins. The antibody recognizes the epitope tag fused to either the amino- or carboxy- termini of targeted proteins.Polyclonal anti-MYC (EQKLISEEDL) reacts with human c- MYC tagged fusion proteins. The antibody recognizes the epitope tag fused to either the amino- or carboxy- termini of targeted proteins. This conjugate is prepared by labeling polyclonal rabbit anti-MYC IgG (H+L) with CF™543 dye. CF™543 is an orange fluorescent dye with an excitation spectrum optimally matching the 543 laser line. Antibody conjugates of the dye are much brighter than those of Alexa Fluor® 546.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.

Type: Tag
Antigen: Myc Tag
Clonality: Polyclonal
Clone:
Conjugation: CF543
Epitope:
Host: Rabbit
Isotype:
Reactivity: MYC tag