- Antibody type:Tag
- Antigen name:FLAG Tag (DYKDDDDK)
- Antigen symbol:FLAG Tag
- Clonality:Polyclonal
- Conjugation:CF® 640R
- Host:Rabbit
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
- Storage temperature:Store at –20 °C.
- Concentration:1 mg/mL
- Purification:Affinity purified
- Size:100 μL
- Pk:100 µl
Polyclonal anti-FLAG (DYKDDDDK) reacts with FLAG tagged fusion proteins. The antibody recognizes the epitope tag fused to either the amino- or carboxy- termini of targeted proteins. This conjugate is prepared by labeling polyclonal rabbit anti-FLAG IgG (H+L) with CF™640R dye. CF™640R (Ex/Em 642/662 nm) is a novel far-red rhodamine-based dye spectrally similar to Cy®5 and Alexa Fluor® 647. CF™640R is much brighter than Cy®5 and at least as bright as Alexa Fluor® 647 when excited at 633 or 635 nm. However, the most important advantage of CF™640R is its exceptional photostability.
Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.
Type: Tag
Antigen: FLAG Tag
Clonality: Polyclonal
Clone:
Conjugation: CF640R
Epitope:
Host: Rabbit
Isotype:
Reactivity: FLAG tag