Order Entry
Italy
Orders LinkContactUsLinkComponent
Immunostainer enhancer, Pierce™

Immunostainer enhancer, Pierce™

Immunostain Enhancer, sufficient for 100 large (~3 cm²) tissue section slides, 20 mL
Catalog # PIER46644
Immunostainer enhancer, Pierce™
Catalog # PIER46644
CAS Number:  
Immunostainer enhancer, Pierce™
Immunostainer enhancer, Pierce™

Immunostain Enhancer, sufficient for 100 large (~3 cm²) tissue section slides, 20 mL

CAS Number:  
Price
562,00 €
Log in for availability
1 * 20 mL
Product Details & Documents

The Pierce™ Immunostain Enhancer is compatible with fluorescent and chromogenic detection and routinely increases both signal intensity and detection sensitivity.

  • Only a fraction of antibody is needed to achieve the same signal as with conventional immunodetection
  • Typically provides 3- to 12-fold increase in signal intensity
  • Sensitivity for improved visualisation of the antigen of interest in cells and tissues
  • Significantly improves signal-to-noise ratio for poor quality and low affinity antibodies
  • Can be used with chromogenic and fluorescent detection methods

It enables significant dilution of primary and secondary antibodies. Dilution 5- to 20-fold beyond the normal has been used successfully; however, the dilution factor varies depending on the antibody, tissue, and antigen.

Specifications

Specifications

  • Pk:
    20 mL
  • Description:
    Immunostain Enhancer, sufficient for 100 large (~3 cm²) tissue section slides
  • Cat. No.:
    PIER46644
  • Storage conditions:
    Upon receipt store at 4˚C

Specifications