

Hydrochloric acid 29 - 31%, NORMATOM® for trace metal analysis
Catalog # 85493.290
Supplier: VWR Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Zn (Zinc):max. 1 ppb
- Zr (Zirconium):max. 0.1 ppb
- Assay:29 - 31 %
- Colouration:max. 10 APHA
- Br (Bromide):max. 10 ppm
- Cl2 (Free Chlorine):max. 0.5 ppm
- NH4 (Ammonium):max. 0.5 ppm
- P (Total Phosphorus):max. 0.01 ppm
- S (Total Sulphur):max. 0.3 ppm
- Ag (Silver):max. 1 ppb
- Al (Aluminium):max. 1 ppb
- As (Arsenic):max. 0.5 ppb
- Au (Gold):max. 0.5 ppb
- B (Boron):max. 1 ppb
- Ba (Barium):max. 0.1 ppb
- Be (Beryllium):max. 0.1 ppb
- Bi (Bismuth):max. 0.1 ppb
- Ca (Calcium):max. 1 ppb
- Cd (Cadmium):max. 0.1 ppb
- Ce (Cerium):max. 0.1 ppb
- Co (Cobalt):max. 0.1 ppb
- Cr (Chromium):max. 0.5 ppb
- Cs (Cesium):max. 0.1 ppb
- Cu (Copper):max. 0.5 ppb
- Dy (Dysprosium):max. 0.1 ppb
- Er (Erbium):max. 0.1 ppb
- Eu (Europium):max. 0.1 ppb
- Fe (Iron):max. 1 ppb
- Ga (Gallium):max. 0.1 ppb
- Gd (Gadolinium):max. 0.1 ppb
- Ge (Germanium):max. 1 ppb
- Hf (Hafnium):max. 0.1 ppb
- Hg (Mercury):max. 0.1 ppb
- Ho (Holmium):max. 0.1 ppb
- In (Indium):max. 0.1 ppb
- K (Potassium):max. 1 ppb
- La (Lanthanum):max. 0.1 ppb
- Li (Lithium):max. 0.1 ppb
- Lu (Lutetium):max. 0.1 ppb
- Mg (Magnesium):max. 0.5 ppb
- Mn (Manganese):max. 0.1 ppb
- Mo (Molybdenum):max. 0.1 ppb
- Na (Sodium):max. 1 ppb
- Nb (Niobium):max. 0.1 ppb
- Nd (Neodymium):max. 0.1 ppb
- Ni (Nickel):max. 0.5 ppb
- Pb (Lead):max. 0.1 ppb
- Pd (Palladium):Information Only
- Pr (Praseodymium):max. 0.1 ppb
- Pt (Platinum):Information Only
- Rb (Rubidium):max. 0.1 ppb
- Re (Rhenium):max. 0.1 ppb
- Rh (Rhodium):max. 0.1 ppb
- Ru (Ruthenium):max. 0.1 ppb
- Sb (Antimony):max. 0.5 ppb
- Sc (Scandium):max. 0.1 ppb
- Se (Selenium):max. 1 ppb
- Sm (Samarium):max. 0.1 ppb
- Sn (Tin):max. 0.5 ppb
- Sr (Strontium):max. 0.1 ppb
- Tb (Terbium):max. 0.1 ppb
- Te (Tellurium):max. 0.1 ppb
- Th (Thorium):max. 0.1 ppb
- Ti (Titanium):max. 0.5 ppb
- TI (Thallium):max. 0.1 ppb
- Tm (Thulium):max. 0.1 ppb
- U (Uranium):max. 0.1 ppb
- V (Vanadium):max. 0.5 ppb
- W (Tungsten):max. 0.1 ppb
- Y (Yttrium):max. 0.1 ppb
- Yb (Ytterbium):max. 0.1 ppb
Specification Test Results
About this item
:
Melting Pt:-30 °C
EINECS:231-595-7
ADR:8,II
:36.46 g/mol
UN:1789
REACH:01-2119484862-27
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324