Streptavidin, hydrazide-activated, Pierce™
Streptavidin, hydrazide-activated, 2 mg
Catalog # PIER21120
Supplier: Thermo Fisher Scientific
Streptavidin, hydrazide-activated, Pierce™
Catalog # PIER21120
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Pierce™ Hydrazide-activated strepavidin conjugate includes recombinant streptavidin in a purified form activated for crosslinking to carbonyl groups in a molecule.
Specifications
Specifications
- Pk:2 mg
- Description:Streptavidin, hydrazide-activated