Order Entry
Italy
ContactUsLinkComponent
Pierce™ Cell-free Protein Expression Kits, 1-Step Human High Yield Maxi IVT Kit
Pierce™ Cell-free Protein Expression Kits, 1-Step Human High Yield Maxi IVT Kit
Catalog # PIER88892
CAS Number:  
undefined
Pierce™ Cell-free Protein Expression Kits, 1-Step Human High Yield Maxi IVT Kit
Catalog # PIER88892
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Pk:
    1 KIT
  • Description:
    Step-1 human high yield mini IVT kit, 2×2 ml reactions
Specifications
About this item
The 1-Step Human High-Yield In Vitro Translation (IVT) Kits enable the expression of functional proteins with 10 to 100 times greater yield per ml than other mammalian IVT systems.
  • High expression with up to 750 µg/ml of expressed protein
  • Low variability between experiments
  • Express high levels of protein with 6 hours to overnight incubation
  • Obtain functionally active proteins, including those containing disulfide bonds
  • Transcription and translation is performed in one reaction step
  • Amenable to incorporation of heavy or unnatural amino acids
The 1-Step High-Yield IVT System uses modified HeLa cell extracts to take advantage of the robust human translation machinery and generate functional full-length proteins. In this system, protein expression is performed in a proprietary dialysis device that allows a continuous supply of nucleotides, amino acids and energy generating substrates into the reaction while removing inhibitors of proteins synthesis. This continuous-exchange cell-free (CECF) system enables protein expression in an overnight incubation of up to 750 µg/ml. The complete mini-scale kit includes all the components required for transcription and translation of a recombinant gene, including an optimised expression vector.
Delivery: Each 1- Step Human High-Yield Maxi IVT Kit contains HeLa lysate (4×500 µl), accessory proteins (4×100 µl), reaction mix (4×200 µl), 5X dialysis buffer (23 ml), positive control DNA (pCFE-GFP - 10 µg), cloning vector (pT7CFE1-NHis-GST-CHA - 10 µg), microdialysis devices (2), and nuclease-free water (3×50 ml).