Caspase 3 (cleaved) colorimetric In-Cell ELISA Kit, multispecies, Pierce™
Catalog # PIER62218
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Target protein:Caspase-3
- Description:Caspase 3 Colorimetric In-Cell ELISA Kit, 96 assays
- Tests per kit:96 assays
Specifications
About this item
Cleaved Caspase 3 Colorimetric In-Cell ELISA kit is a colorimetric microplate assay optimised to measure activated (cleaved) caspase 3 and tubulin in whole cells.
- Whole-cell detection requires no cell lysis, protein extraction or special sample preparation
- Provides better data, higher throughput and less cost than quantitative Western blotting
- For use with standard microplate readers
Delivery: The kit includes rabbit monoclonal anti-(cleaved) Caspase antibody, a mouse monoclonal anti-alpha-tubulin antibody, blocking buffer 50 ml, 20X tris buffered saline 50 ml, Surfact-Amps 20 (10% detergent) 10 ml, Surfact-Amps X-100 (10% detergent) 10 ml, HRP conjugate 0,14 ml, TMB substrate 58 ml, TMB stop solution 55 ml, Janus Green whole-cell stain 50 ml, elution buffer 2×30 ml and thin plate seal assembly.