Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Stigmasterol
Stigmasterol ≥95%, powder
Stigmasterol ≥95%, powder
Stigmasterol ≥95%, powder
ICNA0210296225
:
MP Biomedicals
:
Stigmasterol ≥95%, powder
ICNA0210296225
:
MP Biomedicals
:
Pk:
25 g
Seq:
3308
:
:
412.70 g/mol
:
83-48-7
:
C₂₉H₄₈O
:
MFCD00003630