Calcium hydrogen orthophosphate dihydrate, powder USP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:7789-77-7
:
Formula:HCaO₄P·2H₂O
MDL Number:MFCD00149621
:172.09 g/mol
Merck Index:13,01699
Melting Pt:36 °C
Density:2.44 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-826-1
Specification Test Results
- Assay:98% Min.
Specification Test Results
Frequently Bought Together