
1,3-Di-4-piperidylpropane ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16898-52-5
:
Formula:C₁₃H₂₆N₂
MDL Number:MFCD00038013
:210.37 g/mol
Boiling Pt:327 °C
Melting Pt:63…68 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:240-941-6
:GHS07
Related Information
Frequently Bought Together