Inserisci ordine
Italy
ContactUsLinkComponent
2,2-Dimethyl-1,3-dioxan-5-one 90%, tech.
2,2-Dimethyl-1,3-dioxan-5-one 90%, tech.
Catalog # L20177.03
CAS Number:  
2,2-Dimethyl-1,3-dioxan-5-one 90%, tech.
Catalog # L20177.03
CAS Number:  

Specifications

  • Conf:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :130.14 g/mol
Boiling Pt:40…46 °C (14 mmHg)
Storage Temperature:Congelamento
CAS Number:74181-34-3
Formula:C₆H₁₀O₃
MDL Number:MFCD00671514