Order Entry
Italy
ContactUsLinkComponent
3,3-Difluoropyrrolidine hydrochloride 98%
Irritant
3,3-Difluoropyrrolidine hydrochloride 98%
Catalog # L19780.ME
CAS Number:  
Irritant
3,3-Difluoropyrrolidine hydrochloride 98%
Catalog # L19780.ME
CAS Number:  

Specifications

  • Pk:
    500 mg

Specifications

About this item

 :
 :143.56 g/mol
Melting Pt:130…134 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:163457-23-6
Formula:C₄H₇F₂N
MDL Number:MFCD03788948