2-Chlorotrityl alcohol on polystyrene 0.6 - 1.5 mmol/g, cross-linked with 1% DVB 100-200 mesh
Catalog # L19602.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Storage Temperature:Ambient
MDL Number:MFCD02683514