(R)-Methyl-2-amino-2-phenylacetate hydrochloride 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:19883-41-1
:
Formula:C₉H₁₁NO₂·HCl
MDL Number:MFCD00137487
:201.7 g/mol
Melting Pt:∼ 190 °C (dec.)
Storage Temperature:Ambient
EINECS:243-399-9
Related Information
Frequently Bought Together