
Glyoxal dimethyl acetal 60% in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:51673-84-8
:
Formula:OHCCH(OCH₃)₂
MDL Number:MFCD00134410
:104.11 g/mol
Boiling Pt:100…102 °C
Density:1.150
Flash Pt:102 °C (216 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together