
1,3-Bis(3-aminopropyl)tetramethyldisiloxane 94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2469-55-8
:
Formula:C₁₀H₂₈N₂OSi₂
MDL Number:MFCD00039786
:248.52 g/mol
Boiling Pt:132…139 °C (11 mmHg)
Density:0.897
Flash Pt:91 °C (195 °F)
Storage Temperature:Ambient
EINECS:219-588-7
UN:2735
ADR:8,III
:GHS05
Related Information
Frequently Bought Together