Order Entry
Italy
ContactUsLinkComponent
 
2-tert-Butylpropane-1,3-diol 98%
Catalog # L16927.06
CAS Number:  
2-tert-Butylpropane-1,3-diol 98%
Catalog # L16927.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:2819-05-8
 :
Formula:C₇H₁₆O₂
MDL Number:MFCD01075745
 :132.20 g/mol
Boiling Pt:100…102 °C (2 mmHg)
Melting Pt:54…56 °C
Storage Temperature:Ambient