Order Entry
Italy
ContactUsLinkComponent
5-O-tert-Butyldiphenylsilyl-2,3-O-isopropylidene-α,β-D-ribofuranose
5-O-tert-Butyldiphenylsilyl-2,3-O-isopropylidene-α,β-D-ribofuranose
Catalog # L15989.06
CAS Number:  
5-O-tert-Butyldiphenylsilyl-2,3-O-isopropylidene-α,β-D-ribofuranose
Catalog # L15989.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:141607-35-4
 :
Formula:C₂₄H₃₂O₅Si
 :428.6 g/mol
MDL Number:MFCD01075719