

Sodium bromoacetate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1068-52-6
:
Formula:BrCH₂CO₂Na
MDL Number:MFCD01311832
:160.93 g/mol
Melting Pt:>300 °C
ADR:6.1,III
:GHS06
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together