
N-Methylpyridinium-4-carboxaldehyde benzenesulphonate hydrate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:82228-89-5
:
Formula:C₁₃H₁₃NO₄S·xH₂O
MDL Number:MFCD08061351
:279.32 g/mol (anhy)
Melting Pt:98…100 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together