5-Acetyl-2-amino-4-methylthiazole ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:30748-47-1
:
Formula:C₆H₈N₂OS
MDL Number:MFCD00051952
:156.21 g/mol
Melting Pt:∼ 268 °C (dec.)
:GHS06
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together