
1-Phenyl-4,5-dihydro-1H-pyrazol-3-ylamine ≥98%
Catalog # L06476.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
About this item
CAS Number:3314-35-0
:
Formula:C₉H₁₁N₃
MDL Number:MFCD00051730
:161.21 g/mol
Melting Pt:167…170 °C
EINECS:222-005-9
:GHS07
Storage Temperature:Ambient