
2-Chloro-2,2-difluoroacetamide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:354-28-9
:
Formula:ClCF₂CONH₂
MDL Number:MFCD00039775
Melting Pt:79…84 °C
:129.49 g/mol
EINECS:206-555-7
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together