Order Entry
Italy
ContactUsLinkComponent
 
Gold 95% supplied in 0.1 mM PBS, nanoparticles 30 nm OD1 (Abs. 526 nm)
  J67368.AC
 :  
Gold 95% supplied in 0.1 mM PBS, nanoparticles 30 nm OD1 (Abs. 526 nm)
  J67368.AC
 :  

 

  • Pk:
    25 mL
  • Seq:
    2315

 

 

 :
 :196.97 g/mol
 :Refrigerator
 :7440-57-5
 :Au
 :MFCD00003436